Skip to Content
Merck
All Photos(1)

Key Documents

5.08512

Sigma-Aldrich

SNX-482

Synonym(s):

SNX-482

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C192H274N52O60S7
CAS Number:
Molecular Weight:
4495.00
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available. Contact Technical Service

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

15-30 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

storage temp.

−20°C

SMILES string

S1SCC2NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C3N(CCC3)C(=O)C4NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(CSSC4)NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CN)C(C)C)CC(=O)O)CCCCN)C)CCCN\C(=N\[H])\N)C

InChI

1S/C192H274N52O60S7/c1-91(2)57-114-157(270)207-79-146(257)213-135-85-306-307-86-136-183(296)237-133(83-248)179(292)242-153(95(9)10)187(300)231-124(67-141(195)252)170(283)228-126(69-148(260)261)173(286)229-127(70-149(262)263)174(287)239-138(184(297)240-139(189(302)244-55-30-42-140(244)185(298)217-112(161(274)219-114)41-29-54-202-192(198)199)89-311-308-84-134(212-145(256)78-205-155(268)96(11)209-159(272)110(39-26-27-52-193)214-171(284)128(71-150(264)265)232-186(299)152(94(7)8)241-142(253)73-194)181(294)215-111(40-28-53-201-191(196)197)160(273)223-119(63-102-43-47-106(250)48-44-102)164(277)216-113(51-56-305-14)162(275)222-117(60-99-31-18-15-19-32-99)158(271)206-76-143(254)204-77-144(255)211-136)88-310-309-87-137(238-167(280)120(64-103-45-49-107(251)50-46-103)225-177(290)131(81-246)234-165(278)118(61-100-33-20-16-21-34-100)224-163(276)115(58-92(3)4)221-176(289)130(80-245)236-169(282)123(226-182(135)295)66-105-75-200-90-208-105)180(293)210-97(12)156(269)218-122(65-104-74-203-109-38-25-24-37-108(104)109)168(281)227-125(68-147(258)259)172(285)220-116(59-93(5)6)175(288)243-154(98(13)249)188(301)230-121(62-101-35-22-17-23-36-101)166(279)235-132(82-247)178(291)233-129(190(303)304)72-151(266)267/h15-25,31-38,43-50,74-75,90-98,110-140,152-154,203,245-251H,26-30,39-42,51-73,76-89,193-194H2,1-14H3,(H2,195,252)(H,200,208)(H,204,254)(H,205,268)(H,206,271)(H,207,270)(H,209,272)(H,210,293)(H,211,255)(H,212,256)(H,213,257)(H,214,284)(H,215,294)(H,216,277)(H,217,298)(H,218,269)(H,219,274)(H,220,285)(H,221,289)(H,222,275)(H,223,273)(H,224,276)(H,225,290)(H,226,295)(H,227,281)(H,228,283)(H,229,286)(H,230,301)(H,231,300)(H,232,299)(H,233,291)(H,234,278)(H,235,279)(H,236,282)(H,237,296)(H,238,280)(H,239,287)(H,240,297)(H,241,253)(H,242,292)(H,243,288)(H,258,259)(H,260,261)(H,262,263)(H,264,265)(H,266,267)(H,303,304)(H4,196,197,201)(H4,198,199,202)

InChI key

NSUPRLHDCFNOKD-UHFFFAOYSA-N

General description

A peptidyl toxin present in the venom of African tarantula, Hysterocrates gigas that acts as a high affinity, reversible blocker of Cav2.3 (α1E, R-type) channels (IC50 = 15-30 nM). The inhibition appears to be voltage-dependent. At lower concentrations it is also shown to block native R-type Ca2+ current in rat neurohypophyseal nerve terminals, however, these effects are not observed at higher concentrations. Does not affect Na+ or K+ currents in several cultured cell types (~ 500 nM).

Biochem/physiol Actions

Primary Target
Cav2.3 channels

Warning

Toxicity: Standard Handling (A)

Sequence

GVDKAGCRYMFGGCSVNDDCCPRLGCHSLF→SYCAWDLTFSD (difulfid bond: 7-21, 14-26, 20-33)

Reconstitution

Following reconstitution, aliquot and freeze(-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Abitbol, K. et al. 2012. J. Physiol.590, 2977.
Newcomb, R., et al., 1998. Biochemistry.37, 15353.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karine Abitbol et al.
The Journal of physiology, 590(13), 2977-2994 (2012-05-10)
In the rodent cerebellum, pharmacological activation of mGluR4 acutely depresses excitatory synaptic transmission at parallel fibre–Purkinje cell synapses. This depression involves the inhibition of presynaptic calcium (Ca2+) influx that ultimately controls glutamate release. In this study, we investigate the molecular
R Newcomb et al.
Biochemistry, 37(44), 15353-15362 (1998-11-04)
We describe the first potent and selective blocker of the class E Ca2+channel. SNX-482, a novel 41 amino acid peptide present in the venom of the African tarantula, Hysterocrates gigas, was identified through its ability to inhibit human class E

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service