Skip to Content
Merck
All Photos(1)

Key Documents

5.08410

Sigma-Aldrich

Glycogen Phosphorylase Inhibitor II, KB228

Synonym(s):

Glycogen Phosphorylase Inhibitor II, KB228, KB-228

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N2O7
CAS Number:
Molecular Weight:
354.36
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

930 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A glucopyranosyl urea compound that acts as an inhibitor of muscle glycogen phosphorylase (Ki = 930 nM). Displays a mixed type of inhibition. Enhances glucose sensitivity in chow-fed, obese, diabetic mice and increasing hepatic glucose uptake. Reduces blood glucose levels and increases hepatic glycogen content in C57/BL6J mice. Shown to increase mitochondrial oxidation in Hep2G cells under normoglycemic conditions. Enhances the expression of uncoupling proteins2 (UCP2) in HepG2 cells. This effect was less pronounced under hyperglycemic conditions.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Glycogen Phosphorylase
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Nagy, L., et al. 2013. PLoS One.8, e69420.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service