Skip to Content
Merck
All Photos(1)

Key Documents

5.06419

Sigma-Aldrich

Methysergide Maleate

Synonym(s):

Methysergide Maleate, 5-HT Serotonin Receptor Antagonsit, Methysergide Maleate, UML-491

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$241.00

$241.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$241.00

About This Item

Empirical Formula (Hill Notation):
C21H27N3O2 · xC4H4O4
CAS Number:
Molecular Weight:
353.46 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$241.00


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 10 mM

storage temp.

2-8°C

SMILES string

[n]1(c2c3c(c1)C[C@H]4N(C[C@@H](C=C4c3ccc2)C(=O)N[C@H](CO)CC)C)C.OC(=O)\C=C/C(=O)O

InChI

1S/C21H27N3O2.C4H4O4/c1-4-15(12-25)22-21(26)14-8-17-16-6-5-7-18-20(16)13(10-23(18)2)9-19(17)24(3)11-14;5-3(6)1-2-4(7)8/h5-8,10,14-15,19,25H,4,9,11-12H2,1-3H3,(H,22,26);1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-,15+,19-;/m1./s1

InChI key

LWYXFDXUMVEZKS-ZVFOLQIPSA-N

General description

An antagonist of the 5-HT2B (pki = 8.0-9.4) and 5-HT2C (pki = 8.6-9.1) receptor. At the 5-HT1A and some other 5-HT receptors it serves as a partial agonist. It is known to have partial agonist effects on other 5-HT receptors as well. In neuronal and platelet culture, Methysergide blocks serotonin action with an IC50 = 18 µM. Originally developed in the 1960′s as a migraine/cluster headache treatment, but discontinued due to significant physiological and pyschotropic side effects. Methysergide is still actively used in research as a 5-HT2 receptor antagonist in both in vivo and in vitro experiments.

Biochem/physiol Actions

Primary Target
5-HT

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Matsuta, Y. et al. 2013. Exp. Neurol.247, 250.
IUHPAR Database 2013.

Shad, K.F., Saeed, S.A. 2007. Exp. Brain Res.183, 411.

Knight, A.R. et al. 2004. Naunyn Schmiedebergs Arch. Pharmacol.370, 114.

Saxena, P.R and Lawang, A. 1985. Arch. Int. Pharmacodyn. Ther.277, 235.
Colpaert, F. C. et al. 1979. Eur. J. Pharmacol.58, 505.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service