Skip to Content
Merck
All Photos(1)

Key Documents

506181

Sigma-Aldrich

p75NTR Signaling Inhibitor, Cell-permeable, TAT-Pep5

The p75NTR Signaling Inhibitor, Cell-permeable, TAT-Pep5 controls the biological activity of p75NTR. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

p75NTR Signaling Inhibitor, Cell-permeable, TAT-Pep5, TAT-Pep5, H- YGRKKRRQRRR-CFFRGGFFNHNPRYC-OH, Cyclic

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$431.00

$431.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
$431.00

About This Item

Empirical Formula (Hill Notation):
C150H227N57O32S2
Molecular Weight:
3404.90
UNSPSC Code:
12352202
NACRES:
NA.77

$431.00


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

The p75NTR inhibitor Pep5 (Cat. No. 506180) is made cell-permeable by fusing it with the N-terminal protein transduction domain sequence (11 amino acids) from HIV protein TAT. Shown to act as an effective blocker of MAG- and Nogo-induced inhibition of neurite outgrowth in both DRG (dorsal root ganglion) and postnatal cerebellar neurons. A potential therapeutic agent against inhibition of regeneration in the central nervous system.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Blocker of MAG- and Nogo-induced inhibition of neurite outgrowth
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H₂N-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Cys¹²-Phe-Phe-Arg-Gly-Gly-Phe-Phe-Asn-His-Asn-Pro-Arg-Tyr-Cys²⁶-OH

disulfide bond (12-26)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Yamashita, T., and Tohyama, M. 2003. Nat. Neurosci.6, 461.
Schwarze, S.R., et al. 1999. Science285, 1569.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryo Uetsuki et al.
Biochemistry and biophysics reports, 26, 101003-101003 (2021-05-28)
The low-affinity nerve growth factor receptor p75 is a stratified epithelial stem/progenitor marker of human epithelia. We found OM-1, a human squamous cell carcinoma (SCC) cell line, showed distinct cells with p75 cluster, especially located at the center of a
Wei Sun et al.
Frontiers in cell and developmental biology, 10, 1018586-1018586 (2022-11-29)
Prenatal exposure to maternal infection increases the risk of offspring developing schizophrenia in adulthood. Current theories suggest that the consequences of MIA on mBDNF secretion may underlie the increased risk of cognitive disorder. There is little evidence for whether the
Xinxin Zhou et al.
Molecular pain, 16, 1744806920922100-1744806920922100 (2020-05-02)
HIV-associated neuropathic pain (HNP) is a common complication for AIDS patients. The pathological mechanism governing HNP has not been elucidated, and HNP has no effective analgesic treatment. Brain-derived neurotrophic factor (BDNF) is a member of the neurotrophic factor family related

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service