Skip to Content
Merck
All Photos(1)

Key Documents

506170

Sigma-Aldrich

p53-Snail binding Inhibitor, GN25

The p53-Snail binding Inhibitor, GN25 controls the biological activity of p53-Snail. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

p53-Snail binding Inhibitor, GN25, GN25

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$376.00

$376.00


Estimated to ship on15 May 2025



Select a Size

Change View
25 MG
$376.00

About This Item

Empirical Formula (Hill Notation):
C15H14O6S
Molecular Weight:
322.33
UNSPSC Code:
12352200
NACRES:
NA.77

$376.00


Estimated to ship on15 May 2025


Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, water soluble and specific p53-Snail binding inhibitor that activates p53 in a K-Ras-dependent manner and displays anti-proliferating effect toward K-Ras-MEF (K-Ras-transformed mouse embryonic fibroblast) cells with 20% viability at 10µM, but not in N-Ras/Myc-MEF or non-transformed MEF. Snail and p53 expression are elevated in a dose dependent manner after GN25 treatment at 1-10µM in K-Ras-mutated human cancer cell lines A549, but not in MKN-45 (wt-K-Ras/wt-p53) cells. Furthermore, it is shown to selectively activate wild-type p53 in p53WT/MT cancer cells, but not Panc-1 cells with single allelic mutant p53. Exhibits anti-tumor effect in a xenograft model in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lee, S., et al. 2010. Oncogene29, 4576.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting-Yu Chen et al.
Cell death and differentiation, 28(6), 1865-1879 (2021-01-20)
The DNA-PK maintains cell survival when DNA damage occurs. In addition, aberrant activation of the DNA-PK induces centrosome amplification, suggesting additional roles for this kinase. Here, we showed that the DNA-PK-p53 cascade induced primary cilia formation (ciliogenesis), thus maintaining the
Yu-Ying Chao et al.
Journal of cellular physiology, 237(12), 4487-4503 (2022-10-18)
Pancreatic ductal adenocarcinoma (PDAC) is one of the most lethal cancers because of its late diagnosis and chemoresistance. Primary cilia, the cellular antennae, are observed in most human cells to maintain development and differentiation. Primary cilia are gradually lost during

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service