Skip to Content
Merck
All Photos(1)

Key Documents

5.05417

Sigma-Aldrich

Oxymetazoline Hydrochloride

Synonym(s):

Oxymetazoline Hydrochloride, 6-(tert-butyl)-3-((4,5-dihydro-1H-imidazol-2-yl)methyl)-2,4-dimethylphenol hydrochloride, 3-[(4,5-Dihydro-1H-imidazol-2-yl)me­thyl]-6-(1,1-dimethylethyl)-2,4-dimethyl-phenol hydrochloride, Alpha 2 Adrenergic Receptor Agonist, Oxymetazoline hydrochlo

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$262.00

$262.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$262.00

About This Item

Empirical Formula (Hill Notation):
C16H24N2O · xHCl
CAS Number:
Molecular Weight:
260.37 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$262.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C16H24N2O.ClH/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14;/h8,19H,6-7,9H2,1-5H3,(H,17,18);1H

InChI key

BEEDODBODQVSIM-UHFFFAOYSA-N

General description

α2-Adrenergic receptor agonist. Exhibits over 60-fold greater selectivity for α2A receptors over α receptors. Enhances EGF- and PDGF-induced DNA synthesis.

Biochem/physiol Actions

Primary Target
α₂ adrenergic

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Makara, G.B., et al. 1995. Eur. J. Pharmacol.287, 43.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service