Skip to Content
Merck
All Photos(1)

Key Documents

5.05313

Sigma-Aldrich

Hexamethonium Bromide

Synonym(s):

Hexamethonium Bromide, Hexane-1,6-bis(trimethylammonium bromide), Nicotinic Acetylcholine Receptors (nAChRs) Antagonist, Hexamethonium Bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H30Br2N2
CAS Number:
Molecular Weight:
362.19
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (TLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

H2O: 100 mg/mL

storage temp.

10-30°C

InChI

1S/C12H30N2.2BrH/c1-13(2,3)11-9-7-8-10-12-14(4,5)6;;/h7-12H2,1-6H3;2*1H/q+2;;/p-2

InChI key

FAPSXSAPXXJTOU-UHFFFAOYSA-L

General description

A potent antagonist of nicotinic acetylcholine receptors (nAChRs) at pre-ganglionic sites in both the sympathetic and parasympathetic nervous systems. Often used for studying the roles of nicotinic receptors in regulating cardiovascular system.

Biochem/physiol Actions

Primary Target
nAChR

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Dasgupta, X., et al., 2010. Proc. Natl. Acad. Sci.103, 6332.
Karavaev, N., et al., 2008. Neurosci. Behav. Physiol.38, 297.
Nelson, E., 1999. J. Physiol.516, 657.
Christ, D. 1980. Br. J. Pharmacol.69, 249.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service