Skip to Content
Merck
All Photos(1)

Key Documents

5.04466

Sigma-Aldrich

LY-404,039

Synonym(s):

LY-404,039, (1R,4S,5S,6S)-4-[[(2S)-2-Amino-4-(methylthio)-1-oxobutyl]amino]-2-thiabicyclo[3.1.0]hexane-4,6-dicarboxylic acid 2,2-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO6S
CAS Number:
Molecular Weight:
235.21
UNSPSC Code:
12352200

Pricing and availability is not currently available.

Assay

≥98% (NMR)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 0.25 mg/mL
dilute NaOH: 10 mg/mL

storage temp.

−20°C

InChI

1S/C12H18N2O7S2/c1-22-3-2-5(13)9(15)14-12(11(18)19)4-23(20,21)8-6(7(8)12)10(16)17/h5-8H,2-4,13H2,1H3,(H,14,15)(H,16,17)(H,18,19)/t5-,6+,7+,8-,12+/m0/s1

InChI key

VOYCNOJFAJAILW-OZFRBSTCSA-N

General description

A highly selective and potent agonist for recombinant human mGlu2 and mGlu3 receptors (Ki = 149 nM and 92 nM, respectively) and for rat neurons mGlu2/3 receptors (Ki = 88 nM). Negligible interactions with other mGlu receptors and ionotropic glutamate receptors, glutamate transporter subtypes, monoamine and other receptors. Widely used for investigating neuropsychiatric disorders in animal models and developing potential treatment for human mental disorders, such as schizophrenia.

Biochem/physiol Actions

Primary Target
mGlu₂ & mGlu₃
Target Ki: 149 nM and 92 nM for human mGlu₂ and mGlu₃, respectively.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 3 months ta -20°C.

Other Notes

Seeman, P., 2013. Neuropharmacology.66, 87.
Rorick-Kehn, L., et al. 2007. J. Pharmacol. Exp. Ther.321, 308.
Fell, M., et al. 2008. J. Pharmacol. Exp. Ther.326, 209.
Rodd, Z., et al. 2006. Behav. Brain Res.171, 207.
Mezler, M., et al. 2010. Curr. Opi. In Investigational Drugs.11, 833.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service