Skip to Content
Merck
All Photos(1)

Key Documents

5.00670

Sigma-Aldrich

FATP2 Inhibitor, CB-2

Synonym(s):

FATP2 Inhibitor, CB-2, (5E)-5-((3-bromo-4-hydroxy-5-methoxyphenyl)methylene)-3-(3-chlorophenyl)-2-thioxothiazolidin-4-one, CB2, Fatty Acid Transport Protein 2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5006700001
$244.00

$244.00


Estimated to ship on21 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5006700001
$244.00

About This Item

Empirical Formula (Hill Notation):
C17H11BrClNO3S2
Molecular Weight:
456.76
UNSPSC Code:
41106300
NACRES:
NA.77

$244.00


Estimated to ship on21 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

8 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell permeable thioxothiazolindine compound that reversibly inhibits FATP2-mediated long chain fatty acid uptake in Caco-2 and HepG2, but not non-FATP2-expressing 3T3-L1 adipocytes, cultures (IC50 = 3.99, 7.55, and 231.1 µM, respectively), while exhibiting much reduced potency toward cellular Acsl (acyl-CoA synthetase) activity (IC50 = 50 µM). Shown not to affect Caco-2 viability, membrane permeability, or glucose transport.

Biochem/physiol Actions

Cell permeable: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sandoval, A., et al. 2010. Biochem. Pharmacol.79, 99.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service