Skip to Content
Merck
All Photos(1)

Key Documents

5.00655

Sigma-Aldrich

Rad6 Inhibitor, TZ9

Synonym(s):

Rad6 Inhibitor, TZ9, TZ9, TZ-9, TZ 9, HHR6 Inhibitor, 4-Amino-6-(phenylamino)-[1,3,5]triazin-2-yl)methyl-4-nitrobenzoate, (4-Amino-6-(phenylamino)-1,3,5-triazin-2-yl)methyl 4-nitrobenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$198.00
1.1 G
Inquire

$198.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$198.00
1.1 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C17H14N6O4
CAS Number:
Molecular Weight:
366.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$198.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

pale yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

C1=CC=C(C=C1)NC2=NC(=NC(=N2)N)COC(=O)C3=CC=C(C=C3)[N+](=O)[O-]

InChI

1S/C17H14N6O4/c18-16-20-14(21-17(22-16)19-12-4-2-1-3-5-12)10-27-15(24)11-6-8-13(9-7-11)23(25)26/h1-9H,10H2,(H3,18,19,20,21,22)

InChI key

RRRDZFQRNJTKHL-UHFFFAOYSA-N

General description

A cell-permeable triazine compound that directly targets human E2 Ubiquitin-conjugating enzyme Rad6B/HHR6B catalytic site via noncovalent interactions and effectively inhibits Rad6B-Ub thioester bond formation as well as subsequent substrate ubiquitination without affecting UbcH5B/UBE2D2-catalyzed BCA2 ubiquitination. Shown to inhibit the proliferation of high Rad6B-expressing MDA-MB-231 (IC50 = 6 µM in 72 h), but not the low Rad6B-expressing, non-transformed MCF10A, via G2/M arrest and apoptosis induction. Two known cellular Rad6 substrates, β-catenin and PCNA, are reported to be downregulated following 24 h inhibitor treatment in MDA-MB-231 cultures (0.5 to 5 µM).
A cell-permeable triazine compound that is reported to directly target human E2 Ubiquitin-conjugating enzyme Rad6B/HHR6B catalytic site via noncovalent interactions and effectively inhibit Rad6B-Ub thioester bond formation and subsequent Ub transfer to H2A (by 61%; 1 h 25 nM drug preincubation prior to 1 h H2A ubiquitination reactoin) without affecting UbcH5B/UBE2D2-catalyzed BCA2 ubiquitination. Shown to inhibit the proliferation of high Rad6B-expressing MDA-MB-231 (IC50 = 6 µM in 72 h by MTT assays; 96.3% inhibition of 24 h colony formation by 10 µM inhibitior) via G2/M arrest and apoptosis induction, while exhibiting little toxicity toward low Rad6B-expressing, non-transformed MCF10A (2% and 19% inhibition in 72 h by 10 and 50 µM inhibitor, respectively; by MTT assays). Two known cellular Rad6 substrates, β-catenin and PCNA, are shown to be downregulated following 24 h inhibitor treatment in a dose-dependent manner in MDA-MB-231 cultures (0.5 to 5 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Rad6B/HHR6B
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Sanders, M.A., et al. 2013. Mol. Cancer Ther.12, 373.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service