Skip to Content
Merck
All Photos(1)

Key Documents

5.00507

Sigma-Aldrich

CXCR4 Antagonist IV, TF14016

Synonym(s):

CXCR4 Antagonist IV, TF14016, Fusin Antagonist IV

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$348.00

$348.00


Estimated to ship on23 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
2 MG
$348.00

About This Item

Empirical Formula (Hill Notation):
C101H145FN34O18S2 · xH2O
Molecular Weight:
2206.58 (anhydrous basis)
UNSPSC Code:
51111800
NACRES:
NA.77

$348.00


Estimated to ship on23 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

solid

potency

0.9 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

H2O: 100 mg/mL

storage temp.

−20°C

General description

A 14-aa internally disulfide-bonded peptide that potently competes against SDF-1α/CXCL12 for CXCR4 binding (IC50 = 0.91 nM; [SDF-1] = 100 nM) and protects MT-4 cells against X4-HIV strain HIV-1IIIB infection (EC50 = 4 nM in 5 d; MOI = 0.01) with no significant cytotoxicity (CC50 = 56 µM; 5 d). Inhibits SDF-1-induced Ca2+ mobilization (IC50 = 4.5 nM; [SDF-1] = 30 nM; CXCR4-expressing CHO cells) in vitro and effectively prevents CXCR4-dependent 5BC-5 metastasis in NK-depleted SCID mice in vivo (10 mg/kg i.p.) in vivo.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CXCR4

Warning

Toxicity: Standard Handling (A)

Sequence

Nα-4-fluorobenzoyl-Arg-Arg-Nal-Cys⁴-Tyr-Cit-Lys-D-Lys-Pro-Tyr-Arg-Cit-Cys¹³-Arg-NH₂ (Disulfide bond: 4 → 13; Nal = L-3-(2-naphthyl)alanine; Cit = L-citrulline)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Otani, Y., et al. 2012. FEBS Lett.586, 3639.
Oishi, S., and Fujii, N. 2012. Org. Biomol. Chem.10, 5720.
Tamamura, H., et al. 2003. Org. Biomol. Chem.1 3663.
Tamamura, H., et al. 2003. Org. Biomol. Chem.1, 3656.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service