Skip to Content
Merck
All Photos(1)

Key Documents

5001S

Sigma-Aldrich

Bovine Serum Albumin(BSA) Sampling Kit

29-31 g/dL protein (Biuret), USA Origin, enzyme immunoassay: suitable, Serology: suitable, Blood banking: suitable

Synonym(s):

30% Liquid Sampling Kit

Sign Into View Organizational & Contract Pricing

Select a Size

5001S
$265.00

$265.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
5001S
$265.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801
NACRES:
NA.28

$265.00


Estimated to ship on20 May 2025


Request a Bulk Order

Product Name

Bovine Serum Albumin(BSA) Sampling Kit, 30% BSA, liquid

form

liquid

Quality Level

packaging

pkg of 4 × 50 mL

origin

USA origin

concentration

30% BSA

technique(s)

blood typing: suitable
enzyme immunoassay: suitable

dilution

(Serology)

foreign activity

Bluetongue, none detected
Vesicular Stomatitis Virus, none detected

Looking for similar products? Visit Product Comparison Guide

Components

81070N, 81077N, 81177N, 820464

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service