Skip to Content
Merck
All Photos(1)

Key Documents

492000-M

Millipore

NOX Inhibitor III, VAS2870

The NOX Inhibitor III, VAS2870, also referenced under CAS 722456-31-7, controls the biological activity of NOX.

Synonym(s):

NOX Inhibitor III, VAS2870, 7-(1,3-Benzoxazol-2-ylsulfanyl)-3-benzyl-3H-[1,2,3]triazolo[4,5-d]pyrimidine, 1,3-Benzoxazol-2-yl-3-benzyl-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-yl sulfide, NADPH Oxidase Inhibitor III, VAS2870

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12N6OS
CAS Number:
Molecular Weight:
360.39
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

General description

A cell-permeable thiotriazolopyrimidine compound that acts as an effective NADPH oxidase (NOX) inhibitor (IC50 = 10.6 µM against NOX activity in human neutrophil lysates) and is widely used for studying NOX-dependent cellular ROS formations in various cultures, including oxLDL-treated HUVEC (complete inhibition at 5 µM), PDGF-induced primary rat VSMC (vascular smooth muscle cells; complete inhibition at 10 µM), as well as aortic sections from spontaneously hypertensive rats (72% inhibition at 10 µM) ex vivo.
A cell-permeable thiotriazolopyrimidine compound that acts as an effective NADPH oxidase (NOX) inhibitor (IC50 = 10.6 µM against NOX activity in human neutrophil lysates) and is widely used for studying NOX-dependent cellular ROS formations in various cultures, including oxLDL-treated HUVEC (complete inhibition at 5 µM), PDGF-induced primary rat VSMC (vascular smooth muscle cells; complete inhibition at 10 µM), as well as aortic sections from spontaneously hypertensive rats (SHRs) (72% inhibition at 10 µM). Also shown to inhibit PDGF-BB-induced murine EB (embryonid body) vacularization (74% inhibition at 50 µM) and improve ACh-induced vasorelaxation of isolated rat aortic rings from both SHRs and non-hypertensive rats ex vivo (by 41.7 and 34.5%, respectively, at 10 µM).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wind, S., et al. 2010. Hypertension56, 490.
Lange, S., et al. 2009. Cardiovasc. Res.81, 159.
Stielow, C., et al. 2006. Biochem. Biophys. Res. Commun.344, 200.
ten Freyhaus, H., et al. 2006. Cardiovasc. Res.71, 331.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service