Skip to Content
Merck
All Photos(1)

Documents

4.90289

Sigma-Aldrich

(2-Hydroxyethyl)trimethylammonium dimethylphosphate

for synthesis

Synonym(s):

(2-Hydroxyethyl)trimethylammonium dimethylphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H20NO5P
CAS Number:
Molecular Weight:
229.21
UNSPSC Code:
12352107

Quality Level

form

solid

mp

40-41 °C

storage temp.

2-30°C

Analysis Note

Identity (NMR): passes test
Assay (cations): ≥ 98.0 %
Halides (IC): ≤ 0.1 %
Water (K. F.): ≤ 1.0 %

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service