Skip to Content
Merck
All Photos(1)

Key Documents

487664

Sigma-Aldrich

p-Nitrophenyl Phosphate Substrate Buffer

This p-Nitrophenyl Phosphate Substrate Buffer is validated for use in ELISA.

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$232.00

$232.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$232.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.77

$232.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

sterility

non-sterile

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clearlight yellow

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

Alkaline phosphatase substrate buffer supplied as ready-to-use. A diluent for Cat. No. 487663.
This p-Nitrophenyl Phosphate Substrate Buffer is validated for use in ELISA.

Application

ELISA (see comments)

Biochem/physiol Actions

Primary Target
Alkaline phosphatase substrate buffer
Product does not compete with ATP.

Warning

Toxicity: Standard Handling (A)

Other Notes

Product tested for stability at 4°C, 18-26°C, and 37°C. Product performance is tested on the final product by ELISA.



Suggested procedure for use of p-NPP in alkaline phosphatase-based ELISAs:



1. To 39 parts distilled water add 1 part 50X p-NPP Concentrate (Cat. No. 487663) and 10 parts p-NPP Substrate Buffer. Mix well and store protected from light. This diluted reagent is suitable for up to 10 h after preparation.

2. Complete all required incubations with antibodies and alkaline phosphatase-labeled probes.

3. Wash plate wells at least 4 times with Tris-buffered saline (TBS) containing 0.1% Tween®-20 detergent. DO NOT USE PHOSPHATE BUFFERS; inorganic phosphate is a potent inhibitor of alkaline phosphatase.

4. After the final wash, shake and blot all residual buffer from the wells.

5. Add 100 µl of 1X p-NPP Substrate Solution to each well and incubate for 15-60 min. (If a kinetic study is desired, readings at 405 nm may be taken at intermediate intervals).

[Note: Optimal incubation times may vary depending on the amount of enzyme present. If color develops too quickly, dilution of the probe, antibody, or alkaline phophatase-labeled reagent may be required.]

6. Stop the reaction by adding 100 µl of 1 N NaOH.

7. Read absorbance at 405 nm.



Note: Variations in time, reagent volumes, and temperature may require further standardization by the user.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service