Skip to Content
Merck
All Photos(1)

Key Documents

481925

Sigma-Aldrich

NADH Oxidase, Bacillus licheniformis

Native NADH oxidase from Bacillus Iicheniformis. Catalyzes the oxidation of NADPH to NAD and H2O2 with the consumption of stoichiometric amounts of oxygen.

Synonym(s):

NADH Oxidase, Bacillus licheniformis, Nicotinamide-adenine Dinucleotide Oxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

Quality Level

form

lyophilized

specific activity

≥50 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

Native NADH oxidase from Bacillus Iicheniformis. Catalyzes the oxidation of NADPH to NAD and H2O2 with the consumption of stoichiometric amounts of oxygen.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will oxidize 1 µmol of NADH per min at 30°C, pH 7.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or store at room temperature (20°C) for short-term storage. Stock solutions are stable for up to 1 week at 20°C or for up to 6 months at -20°C.
Reconstitute to 5 units/ml in 30 mM potassium phosphate buffer, pH 7.4. Dilute in 30 mM potassium phosphate buffer, pH 7.4, with 0.1% BSA just prior to use.

Other Notes

May contain trace amounts of catalase which can be eliminated by the addition of 10 mM NaN₃ to the reaction mixture.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service