Skip to Content
Merck
All Photos(1)

Key Documents

481909

Sigma-Aldrich

Niclosamide

A salicylanilide anthelmintic drug that acts as a potent and selective Stat3- and Wnt-signaling pathway inhibitor.

Synonym(s):

Niclosamide, 5-Chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxy-benzamide, mTOR Inhibitor IX, Autophagy Inducer III, STAT3 Inhibitor XV, Wnt Pathway Inhibitor XIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8Cl2N2O4
CAS Number:
Molecular Weight:
327.12
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 2.5 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C13H8Cl2N2O4/c14-7-1-4-12(18)9(5-7)13(19)16-11-3-2-8(17(20)21)6-10(11)15/h1-6,18H,(H,16,19)

InChI key

RJMUSRYZPJIFPJ-UHFFFAOYSA-N

General description

A cell-permeable salicylanilide that, in addition to its well-known antihelmintic efficacy and use in tapeworm treatment, acts as a mammalian mTORC1, but not mTORC2, signaling inhibitor mechanistically distinct from rapamycin, causing MCF-7 autophagy activation under nutrient-rich conditions (by 15-fold with 4h 10 M treatment), SH-SY5Y lysosome redistribution (10 M for 8 h), and dissipation/clearance of ubiquinated protein aggregates upon proteasome inhibition in a reversible, time- and dose-dependent manner. Likely a direct consequence of autophagy activation, Niclosamide is demonstrated to induce Wnt-independent Frizzled1 endocytosis (t1/2 = 2.4 h; effective conc. <2 M) and Dishevelled-2 downregulation (IC50 ~1 M), effectively preventing Wnt3a from stimulating HEK293 cellular LEF/TCF transcription activity (IC50 = 0.5 M TOPFlash reporter assays). Unrelated to its autophagy induction activity, Niclosamide is also shown to inhibit Stat3 signaling (IC50 = 0.25 M in HeLa-based pLucTKS3 reporter assays) by suppressing the phosphorylation of Jak1 Tyr1022/1023 (2 h treatment at 0.5 to 2.0 M in Du145 cultures) and Stat3 Tyr705 (by >90% after 24 h treatment at 2 and 5 M, respectively, in Du145 and HeLa cultures), without affecting the phosphorylation of Stat1 Tyr701, Stat3 Ser727, Stat5 Tyr694, or Jak2 Tyr1007/1008.
A cell-permeable salicylanilide that, in addition to its well-known antihelmintic efficacy, acts as a mammalian mTORC1, but not mTORC2, signaling inhibitor mechanistically distinct from rapamycin. Likely a direct consequence of autophagy activation, Niclosamide is demonstrated to induce Wnt-independent Frizzled1 and Dishevelled-2 downregulation. Unrelated to its autophagy induction activity, Niclosamide is also shown to inhibit Stat3 signaling (IC50 = 0.25 M in HeLa reporter assays). Efficiently inhibits breast cancer stem-like cells in vitro and in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wang, Y.C., et al. 2013. PLoS ONE8, e74538.
Chen, W., et al. 2010. Am. J. Physiol. Gastrointest. Liver Physiol.299, G293.
Gies, E., et al. 2010. PLoS ONE5, e14410.
Ren, X., et al. 2010. ACS Med. Chem. Lett.1, 454.
Chen, M., et al. 2009. Biochemistry48, 10267.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiuge Tang et al.
International journal of molecular sciences, 25(8) (2024-04-27)
Canonical autophagy is an evolutionarily conserved process that forms double-membrane structures and mediates the degradation of long-lived proteins (LLPs). Noncanonical autophagy (NCA) is an important alternative pathway involving the formation of microtubule-associated protein 1 light chain 3 (LC3)-positive structures that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service