Skip to Content
Merck
All Photos(1)

Key Documents

480402

Sigma-Aldrich

NFAT Inhibitor

The NFAT Inhibitor controls the biological activity of NFAT. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

NFAT Inhibitor, MAGPHPVIVITGPHEE

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$218.00

$218.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
$218.00

About This Item

Empirical Formula (Hill Notation):
C75H118N20O22S
CAS Number:
Molecular Weight:
1683.93
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$218.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

S(CC[C@H](N)C(=O)N[C@@H](C)C(=O)NCC(=O)N1[C@@H](CCC1)C(=O)N[C@@H](Cc5nc[nH]c5)C(=O)N2[C@@H](CCC2)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H]([C@H](O)C)C(=O)NCC(=O)N3[C@@H](CCC3)C(=O)N[C@@H](Cc4nc[nH]c

InChI

1S/C75H118N20O22S/c1-12-39(7)59(90-70(111)57(37(3)4)88-68(109)52-19-16-27-95(52)74(115)49(30-44-32-78-36-82-44)87-67(108)51-18-15-26-94(51)53(97)33-79-62(103)41(9)83-63(104)45(76)24-28-118-11)72(113)89-58(38(5)6)71(112)91-60(40(8)13-2)73(114)92-61(42(10)9

InChI key

QPMHUXBSHGAVGD-MCDIZDEASA-N

General description

A highly selective NFAT (Nuclear Factor of Activated T-cells) inhibitor that interferes with calcineurin-NFAT interaction without affecting calcineurin (Cat. No. 539568) phosphatase activity. Inhibits NFAT activation and NFAT-dependent expression of endogenous cytokine genes in T cells, without affecting the expression of other cytokines that require calcineurin but not NFAT.
A highly selective NFAT inhibitor that prevents NFAT activation and NFAT-dependent expression of endogenous cytokine genes in T cells. Does not affect the expression of other cytokines requiring calcineurin, but not NFAT. Inhibits calcineurin-mediated dephosphorylation of NFAT1, NFAT2, and NFAT4 in cell extracts.

Biochem/physiol Actions

Cell permeable: no
Primary Target
NFAT (Nuclear Factor of Activated T-cells)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Met-Ala-Gly-Pro-His-Pro-Val-Ile-Val-Ile-Thr-Gly-Pro-His-Glu-Glu-OH

Other Notes

Noguchi, H., et al. 2004. Nat. Med.10, 305.
Kiani, A., et al. 2001. Blood98, 1480.
Aramburu, J., et al. 1999. Science285, 2129.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sami I Said
Annals of the New York Academy of Sciences, 1144, 148-153 (2008-12-17)
Pulmonary vascular remodeling and inflammation often coexist in clinical and experimentally induced pulmonary arterial hypertension (PAH). In some instances, the pulmonary hypertension may be the primary, or at least the initial, problem, while inflammatory or autoimmune responses appear to initiate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service