Skip to Content
Merck
All Photos(1)

Key Documents

4.80170

Supelco

Trifluoroacetic acid solution

0.05% (v:v) trifluoroacetic acid for liquid chromatography LiChrosolv®

Synonym(s):

Water with 0.05% (v:v) trifluoroacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
$134.00

$134.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
$134.00

About This Item

UNSPSC Code:
12352401
NACRES:
NA.03
technique(s):
HPLC: suitable

$134.00


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

description

isocratic

product line

LiChrosolv®

form

liquid

concentration

0.045-0.055 % (v/v)

technique(s)

HPLC: suitable

density

1.000 g/cm3 at 20 °C

storage temp.

2-30°C

General description

LiChrosolv hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionization methods (ESI/APCI - positive and negative mode). Thanks to their low level of ionic background and low ion suppression, LiChrosolv hypergrade solvents provide high reproducibility and high ionization efficiency. The packaging material has been improved to meet LC-MS quality requirements perfectly.

Analysis Note

Assay (trifluoroacetic acid): 0.045 - 0.055 % (v/v)

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service