Skip to Content
Merck
All Photos(1)

Key Documents

480030

Sigma-Aldrich

NEMO-Binding Domain Binding Peptide, Cell-Permeable, Negative Control

A cell-permeable Antennapedia-NBD mutated fusion peptide analog of NEMO-Binding Domain Binding Peptide that serves as a negative control.

Synonym(s):

NEMO-Binding Domain Binding Peptide, Cell-Permeable, Negative Control, DRQIKIWFQNRRMKWKK-TALDASALQTE, NBD-Binding Peptide, Cell-Permeable, Negative Control

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$612.00

$612.00


Estimated to ship on19 May 2025



Select a Size

Change View
500 μG
$612.00

About This Item

Empirical Formula (Hill Notation):
C154H249N47O42S
Molecular Weight:
3462.98
UNSPSC Code:
12352200
NACRES:
NA.77

$612.00


Estimated to ship on19 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable Antennapedia-NBD mutated (Trp739 and Trp741 replaced with Ala) fusion peptide analog of NEMO-Binding Domain Binding Peptide (Cat. No. 480025) that serves as a negative control. Shown to be defective in binding to NEMO. Does not affect the TNF-α-induced phosphorylation of c-Jun and DNA binding of the transcription factor Oct-1.
A cell-permeable, Antennapedia-NBD mutated (Trp739 → Ala and Trp741 → Ala) fusion peptide analog of NEMO-Binding Domain Binding peptide (Cat. No. 480025) that serves as a negative control. Reported to be defective in binding to NEMO. Does not affect the TNF-α-induced phosphorylation of c-Jun or DNA binding of the transcription factor Oct-1.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Negative control defective in binding to NEMO
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H₂N-Asp-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Thr-Ala-Leu-Asp-Ala-Ser-Ala-Leu-Gln-Thr-Glu-OH

Other Notes

Jimi, E., et al. 2004. Nat. Med., 10, 617.
May, M.J., et al. 2000. Science289, 1550.
This peptide will precipitate when added to tissue culture medium even at 100 µM concentrations; however, sufficient material will stay in solution to permeate cells.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service