Skip to Content
Merck
All Photos(1)

Documents

480026

Sigma-Aldrich

NBD-Binding Peptide II, Cell Permeable, CTP-NBD

A cell-permeable cytoplasmic transduction peptide (CTP; YGRRARRRARR) fused to the NEMO binding domain peptide (NBD; TALDWSWLQTE) that acts as a potent inhibitor of TNFα-stimulated NF-κB activity (IC₅₀~ 50 µM in HCT116 cells).

Synonym(s):

NBD-Binding Peptide II, Cell Permeable, CTP-NBD, NEMO-Binding Domain Binding Peptide II, Cell-Permeable, CTP-NBD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C123H197N47O32
Molecular Weight:
2846.18
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable cytoplasmic transduction peptide (CTP; YGRRARRRARR) fused to the NEMO binding domain peptide (NBD; TALDWSWLQTE) that acts as a potent inhibitor of TNFα-stimulated NF-κB activity (IC50 ~ 50 µM in HCT116 cells). Acts by interfering with the formation of active IKK complex and thereby prevents IκBα phosphorylation and its subsequent proteasomal degradation. Also blocks LPS-mediated increase in inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) activities. Colon-targeted capsules containing CTP-NBD are shown to reduce 2,4,6-trinitrobenzenesulfonic acid (TNBS)-induced colitis in rodent models (4.5 mg/kg/,p.o.) in vivo and diminish activities of myeloperoxidase and several inflammatory mediators. Antennapedia fused NBD cell-permeable peptide is also available (>Cat. No. 480025).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NEMO-binding peptide
Reversible: yes

Packaging

Packaged under inert gas
Yes

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-YGRRARRRARRTALDWSWLQTE-NH₂

Other Notes

Hong, S., et al. 2012. Mol. Pharm.9, 1310.
Kim, D., et al. 2006. Exp. Cell Res.312, 1277.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service