Skip to Content
Merck
All Photos(1)

Key Documents

475904-M

Millipore

MOWIOL® 4-88 Reagent

Synonym(s):

Poly(vinyl alcohol)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$195.00

$195.00


Estimated to ship on25 May 2025



Select a Size

Change View
100 G
$195.00

About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
41202101
NACRES:
NA.75

$195.00


Estimated to ship on25 May 2025


description

RTECS - TR8100000

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

storage temp.

10-30°C

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Used for mounting cells on slides for detection of cell-surface antigens by immunofluorescence. Useful as an anti-fading agent.

Warning

Toxicity: Standard Handling (A)

Preparation Note

Add 2.4 g of Mowiol® 4-88 to 6 g of glycerol and stir to mix. Add 6 ml of water and stir for several hours at room temperature. Add 12 ml of 0.2 M Tris (pH 8.5) and heat to 50°C for 10 min. with occasional mixing. After the Mowiol 4-88 dissolves, clarify the solution by centrifugation at 5000 x g for 15 min.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 2 weeks at 4°C.

Other Notes

Longin, A., et al. 1993. J. Histochem. Cytochem. 41, 1833.
Beug, H., et al. 1979. Cell18, 375.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Mowiol is a registered trademark of Kuraray Europe GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service