Skip to Content
Merck
All Photos(1)

Key Documents

454575

Sigma-Aldrich

N-Methyl-D-aspartic Acid hydrate

>95% (TLC), solid, excitatory amino acid neurotransmitter, Calbiochem®

Synonym(s):

N-Methyl-D-aspartic Acid, Hydrate, NMDA

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$228.00

$228.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$228.00

About This Item

Empirical Formula (Hill Notation):
C5H9NO4 · xH2O
CAS Number:
Molecular Weight:
147.13 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77

$228.00


Estimated to ship on26 May 2025


Request a Bulk Order

Product Name

N-Methyl-D-aspartic Acid, Hydrate, Excitatory amino acid neurotransmitter.

Quality Level

description

Merck USA index - 14, 6662

Assay

>95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

N([C@H](CC(=O)O)C(=O)[O-])C

InChI

1S/C5H9NO4/c1-6-3(5(9)10)2-4(7)8/h3,6H,2H2,1H3,(H,7,8)(H,9,10)/p-1/t3-/m1/s1

InChI key

HOKKHZGPKSLGJE-GSVOUGTGSA-M

General description

An excitatory amino acid neurotransmitter. Selective agonist of the glutamate receptor that regulates Ca2+ channels. Important in long-term potentiation, ischemia, and epilepsy. NMDA receptors are involved in the "fine tuning" of synaptic connections in the developing brain. Chronic treatment with NMDA produces structural changes in synaptic morphology. Over-excitation of NMDA receptors causes neuronal degeneration and cell death.
Excitatory amino acid neurotransmitter. Selective agonist of the glutamate receptor that regulates Ca2+ channels. Important in long-term potentiation, ischemia, and epilepsy. NMDA receptors are involved in the "fine tuning" of synaptic connections in the developing brain. Chronic treatment with NMDA produces structural changes in synaptic morphology. Over-excitation of NMDA receptors causes neuronal degeneration and cell death.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Glutamate receptor that regulates Ca2+ channels
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for several days at 4°C.

Other Notes

Mattson, M.P., et al. 1993. J. Neurosci. 13, 4575.
Yen, L-H., et al. 1993. J. Neurosci. 13, 4949.
Swann, J.W., et al. 1992. Epilepsy Res. Suppl. 9, 115.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinzheng Guo et al.
STAR protocols, 2(4), 100932-100932 (2021-11-23)
The degeneration of retinal ganglion cells (RGCs) leads to irreversible vision loss in a variety of pathological states. Here, we describe a protocol to evaluate the role of a gene in protecting mouse RGCs when they sustain injuries from excitotoxicity

Questions

  1. What's the difference between CAS Number: 6384-92-5 and CAS Number:454575? (They are both NMDA). Which are applicable to be used in a NMDA lesion surgery?

    1 answer
    1. These products are for research use only and have not been evaluated for in vivo applications. To assess potential differences between this product and M3262, it is recommended to consult the Certificates of Analysis: https://www.sigmaaldrich.com/product/mm/454575#product-documentation and https://www.sigmaaldrich.com/product/sigma/m3262#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service