Skip to Content
Merck
All Photos(2)

Key Documents

444201

Millipore

D-(+)-Melezitose monohydrate

A substrate for α-glucosidase.

Synonym(s):

GlcαFruβ2,1Glcα, H₂O, O-α-D-Glucopyranosyl-(1→3)-β-D-fructofuranosyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$813.00

$813.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$813.00

About This Item

Empirical Formula (Hill Notation):
C18H32O16 · H2O
CAS Number:
Molecular Weight:
522.45
Beilstein:
99539
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

$813.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 5819

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

O.OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](CO)O[C@@]2(CO)O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H32O16.H2O/c19-1-5-8(23)11(26)13(28)16(30-5)32-15-10(25)7(3-21)33-18(15,4-22)34-17-14(29)12(27)9(24)6(2-20)31-17;/h5-17,19-29H,1-4H2;1H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15+,16-,17-,18+;/m1./s1

InChI key

CFXAVQUXSYFPDE-ZDVKUAOASA-N

Looking for similar products? Visit Product Comparison Guide

General description

A substrate for α-galactosidase.
A substrate for α-glucosidase.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tatevossian, A. 1982. Arch. Oral Biol. 27, 39.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service