Skip to Content
Merck
All Photos(1)

Key Documents

438193

Sigma-Aldrich

LRRK2-IN-1

LRRK2-IN-1 primarily used in Inhibition.

Synonym(s):

LRRK2-IN-1, LRRK2 Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$232.00

$232.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$232.00

About This Item

Empirical Formula (Hill Notation):
C31H38N8O3
CAS Number:
Molecular Weight:
570.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$232.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-white to pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)C2CCN(CC2)C(=O)C3=CC(=C(C=C3)NC4=NC=C5C(=N4)N(C6=CC=CC=C6C(=O)N5C)C)OC

InChI key

IWMCPJZTADUIFX-UHFFFAOYSA-N

General description

A cell-permeable, ATP competitive, potent, and selective LRRK2 inhibitor (IC50 of 13 nM, 6 nM, and 2.45 µM for wild type, G2019S mutant, and drug resistant A2016T mutant LRRK2, respectively, in an in vitro ATP-site competititon binding assay). While it is shown to inhibit DCLK2 (IC50 = 45 nM) and suppress MAPK7 autophosphorylation (EC50 = 160 nM), this compound (<10 µM) demonstrates very good overall selectivity, targeting 12 out of 442 other kinases in a kinase-binding and biochemical assay. At 0.05-3 µM, it induces dose-dependent inhibition of Ser910 and Ser935 phosphorylation accompanied by loss of 14-3-3 binding to both wild-type LRRK2 and LRRK2[G2019S] in stably transfected HEK293 cells, but not in the drug-resistant LRRK2[A2016T] and LRRK2[A2016T + G2019S] mutants. Similar effects on endogenous LRRK2 phosphorylation and 14-3-3 binding can be observed in human lympho-blastoid cells and for the LRRK2 G2019S mutant derived from a Parkinson′s disease patient, and in human-derived neuroblastoma SHSY5Y cells, as well as in mouse Swiss 3T3 cells. 100 mg/kg of compound injected into mice elicits complete Ser910 and Ser935 dephosphorylation of LRRK2 in the kidney, but not in the brain which may result from an inability to penetrate the blood-brain barrier. In addition, it promotes relocalization of LRRK2 to more aggregate and fibrillar-like structures.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freexe (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Deng, X., et al. 2011. Nat. Chem. Biol.7, 203.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service