Skip to Content
Merck
All Photos(1)

Key Documents

437996

Sigma-Aldrich

5-Lipoxygenase, Human, Recombinant, S. fruigiperda

5-Lipoxygenase, Human, Recombinant, S. frugiperda E.C 1.13.11.34, catalyzes the formation of 5(S)-HpETE from arachidonic acid and its subsequent conversion to leukotriene A4.

Synonym(s):

5-LO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

form

liquid

specific activity

~20 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human 5-Lipoxygenase expressed in S. fruigiperda insect cells using a baculovirus overexpression system. 5-LO catalyzes the formation of 5(S)-HpETE from arachidonic acid and its subsequent conversion to leukotriene A4. It is localized in the cytosol of resting human and rat peripheral blood neutrophils, whereas it is found in the nucleus in rat basophilic leukemia cells and human alveolar macrophages. Upon stimulation of the cell, 5-LO translocates to the nuclear membrane where it is found in association with 5-LO activating protein (FLAP) and with the 85 kDa cPLA2.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that consumes 1.0 nmol of oxygen per min at 25°C in 50 mM Tris-HCl, 100 µM arachidonate, 2 mM CaCl₂, 1 mM ATP, 250 µM oxygen. Oxygen consumption can be measured using a oxygraph equipped with a Clark oxygen electrode.

Physical form

In 100 mM Tris containing 5 mM EGTA, pH 8.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Brock, T.G., et al. 1997. J. Biol. Chem.272, 8276.
Pouliot, M., et al. 1996. Eur. J. Biochem.238, 250.
Zhang, Y.Y., et al. 1993. J. Biol. Chem.268, 2535.
Shimizu, T., et al. 1984. Proc. Natl. Acad. Sci. USA81, 689.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ha Thi Nguyen et al.
Scientific reports, 10(1), 15965-15965 (2020-10-01)
Natural metabolites with their specific bioactivities are being considered as a potential source of materials for pharmacological studies. In this study, we successfully isolated and identified five known clerodane diterpenes, namely 16-oxo-cleroda-3,13(14)E-dien-15-oic acid (1), 16-hydroxy-cleroda-3,13-dien-15-oic acid (2), 16-hydroxy-cleroda-4(18),13-dien-16,15-olide (3), 3α,16α-dihydroxy-cleroda-4(18),13(14)Z-dien-15,16-olide
Mater H Mahnashi et al.
Evidence-based complementary and alternative medicine : eCAM, 2022, 7921408-7921408 (2022-04-12)
Background. The current study aims to give a scientific origin for employing Habenaria plantaginea Lindl. as a potential candidate against nociception, inflammation, and pyrexia. The pharmacological studies were performed on crude extract and subfractions. In the gas chromatography-mass spectroscopy analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service