Skip to Content
Merck
All Photos(2)

Key Documents

435930

Sigma-Aldrich

LIM Kinase Inhibitor I, LIMKi 3

The LIM Kinase Inhibitor I, LIMKi 3 controls the biological activity of LIM Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

LIM Kinase Inhibitor I, LIMKi 3, N-(5-(1-(2,6-Dichlorophenyl)-3-(difluoromethyl)-1H-pyrazol-5-yl)thiazol-2-yl)isobutyramide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$398.00

$398.00


Estimated to ship on19 May 2025



Select a Size

Change View
10 MG
$398.00

About This Item

Empirical Formula (Hill Notation):
C17H14Cl2F2N4OS
CAS Number:
Molecular Weight:
431.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$398.00


Estimated to ship on19 May 2025


Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC1=CC=CC(Cl)=C1N2N=C(C=C2C3=CN=C(NC(C(C)C)=O)S3)C(F)F

InChI

1S/C17H14Cl2F2N4OS/c1-8(2)16(26)23-17-22-7-13(27-17)12-6-11(15(20)21)24-25(12)14-9(18)4-3-5-10(14)19/h3-8,15H,1-2H3,(H,22,23,26)

InChI key

IVUGBSGLHRJSSP-UHFFFAOYSA-N

General description

A cell-permeable pyrazolylthiazolo-isobutyramide compound that acts as a potent LIM kinase inhibitor (IC50 = 7 and 8 nM against LIMK1 and LIMK2, respectively) and effectively suppresses cellular cofilin phosphorylation (IC50 ~ 1 µM in A549 and MDA-MB-231 cultures) without affecting tubulin polymerization or inducing cytotoxicity (EC50 >10 µM in A549 proliferation and colony formation assays). Shown to effectively destabilize F-actin structure in MDA-MB-231 breast cancer cells (3 to 10 µM) with concomitant blockage of invadopedia-mediated ECM degradation (13% and 10% of control, respectively, by 3 and 10 µM inhibitor) and invasion (45% and 7% of control invasion rate, respectively, by 3 and 10 µM inhibitor). Although reported to exhibit affinity toward AMPKα1 and AMPKα2 in T7 phage-based competition binding studies, inhibitory activity of LIMKi 3 against AMPKα1/2 is not yet directly demonstrated.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LIMK1 and LIMK2
Reversible: yes
Secondary Target
AMPKA1, AMPKA2, DDR1, PAK3, DCAMKL2
Target IC50: 7 and 8 nM against LIMK1 and LIMK2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Scott, R.W., et al. 2010. J. Cell. Biol.191, 169.

Ross-Macdonald, P., et al. 2008. Mol. Cancer Ther.7, 3490.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariana Morales-Quinones et al.
Hypertension (Dallas, Tex. : 1979), 76(2), 393-403 (2020-07-01)
Increased arterial stiffness and vascular remodeling precede and are consequences of hypertension. They also contribute to the development and progression of life-threatening cardiovascular diseases. Yet, there are currently no agents specifically aimed at preventing or treating arterial stiffening and remodeling.
Thomas J Jurrissen et al.
American journal of physiology. Heart and circulatory physiology, 323(5), H879-H891 (2022-09-10)
Adropin is a peptide largely secreted by the liver and known to regulate energy homeostasis; however, it also exerts cardiovascular effects. Herein, we tested the hypothesis that low circulating levels of adropin in obesity and type 2 diabetes (T2D) contribute
Catarina Domingues et al.
Frontiers in cell and developmental biology, 8, 678-678 (2020-09-10)
The mechanical properties of the extracellular environment are interrogated by cells and integrated through mechanotransduction. Many cellular processes depend on actomyosin-dependent contractility, which is influenced by the microenvironment's stiffness. Here, we explored the influence of substrate stiffness on the proteome
Xing Duan et al.
Journal of cellular physiology, 233(8), 6088-6097 (2018-01-11)
LIM kinases (LIMK1/2) are LIM domain-containing serine/threonine/tyrosine kinases that mediate multiple cellular processes in mitosis. In the present study, we explored the functional roles and potential signaling pathway of LIMK1/2 during mouse oocyte meiosis. Disruption of LIMK1/2 activity and expression
Tobias Heib et al.
Cell reports, 35(6), 109102-109102 (2021-05-13)
Megakaryocytes (MKs), the precursors of blood platelets, are large, polyploid cells residing mainly in the bone marrow. We have previously shown that balanced signaling of the Rho GTPases RhoA and Cdc42 is critical for correct MK localization at bone marrow

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service