Skip to Content
Merck
All Photos(1)

Key Documents

420112

Sigma-Aldrich

JAK3 Inhibitor, Negative Control - CAS 153437-07-1 - Calbiochem

The JAK3 Inhibitor, Negative Control, also referenced under CAS 153437-07-1, controls the biological activity of JAK3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JAK3 Inhibitor, Negative Control - CAS 153437-07-1 - Calbiochem, 4-Phenylamino-6,7-dimethoxyquinazoline, HCl, WHI-P258, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O2 · xHCl
CAS Number:
Molecular Weight:
281.31 (free base basis)
UNSPSC Code:
12352200

Quality Level

Assay

≥92% (HPLC)

form

solid

potency

<300 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A useful negative control for JAK3 Inhibitors (Cat. Nos. 420101, 420104, and 420106; IC50 >300 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A negative control for JAK3 inhibitors
Product competes with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 year at 4°C.

Other Notes

Sudbeck, E.A., et al. 1999. Clin. Cancer Res. 5, 1569.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service