Skip to Content
Merck
All Photos(2)

Documents

407325

Sigma-Aldrich

Interferon-α-IFNα-R Interaction Inhibitor

The Interferon-α-IFNα-R Interaction Inhibitor controls the biological activity of Interferon-α-IFNα-R.

Synonym(s):

Interferon-α-IFNα-R Interaction Inhibitor, IFN-α Inhibitor, N-Methyl-1-(2-(naphthalen-1-ylthio)phenyl)methanamine, HCl, N-Methyl-N-(2-(1-naphthylthio)benzyl)amine, HCl, N-Methyl-1-(2-(naphthalen-1-ylthio)phenyl)methanamine, HCl, N-Methyl-N-(2-(1-naphthylthio)benzyl)amine, HCl, IFN-α Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NS · xHCl
Molecular Weight:
279.40 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

CNCC1=CC=CC=C1SC2=CC=CC3=C2C=CC=C3.[xHCl]

General description

A nonpeptidic thiophenylmethanamine compound that binds interferon-α (Kd = 4 µM for human IFN-α) and prevents IFN-α and IFNAR interaction-dependent IFN-α production in murine BM-pDCs (bone marrow-derived plasmacytoid dendritic cells) cultures upon MVA (vaccinia virus Ankara) infection (IC50<4.5 µM), TLR9 ligand CpG2216 stimulation, VSV-M2 (vesicular stomatitis virus variant 2) infection, or MDA5/TLR3 ligand poly(I:C) stimulation (by >95% inhibition at 9 µM), while exhibiting no inhibitory effect against IL-12 production in the same cultures when applied at non-toxic concentrations (≤4.5 µM).
A nonpeptidic thiophenylmethanamine compound that binds interferon-α (Kd = 4 µM for human IFN-α) and prevents IFN-α and IFNAR interaction-dependent IFN-α production in murine BM-pDCs (bone marrow-derived plasmacytoid dendritic cells) cultures upon MVA infection (IC50<4.5 µM), while exhibiting no inhibitory effect against IL-12 production in the same cultures when applied at non-toxic concentrations (≤4.5 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Geppert, T., et al. 2011. Angew. Chem. Int. Ed.51, 258.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debasis Nayak et al.
iScience, 25(5), 104193-104193 (2022-04-29)
The involvement of membrane-bound solute carriers (SLCs) in neoplastic transdifferentiation processes is poorly defined. Here, we examined changes in the SLC landscape during epithelial-mesenchymal transition (EMT) of pancreatic cancer cells. We show that two SLCs from the organic anion/cation transporter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service