Skip to Content
Merck
All Photos(1)

Key Documents

407295

Sigma-Aldrich

Anti-α-Interferon Receptor Chain 2 Mouse mAb (MMHAR-2)

liquid, clone MMHAR-2, Calbiochem®

Synonym(s):

Anti-CD118

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$825.00

$825.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$825.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$825.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MMHAR-2, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... IFNA1(3439)

General description

Purified mouse monoclonal antibody. Recognizes the IFN-α receptor chain 2 protein.
Recognizes the IFN-α receptor chain 2 protein.
This Anti-alpha-Interferon Receptor Chain 2 Mouse mAb is validated for use in FC, IP, Neutralization Studies, Frozen and Paraffin Sections for the detection of α-Interferon Receptor Chain 2.

Immunogen

Human
human α-interferon receptor chain 2

Application

Flow Cytometry (1 µg/10 ml, see comments)

Immunoprecipitation (1-5 µg/ml)

Neutralization Studies (5 µg/ml)

Frozen Sections (1 µg/ml)

Paraffin Sections (1 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 0.1% BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Colamonici, O.R. and Domanski, P. 1993. J. Biol. Chem.268, 10895.
For flow cytometry, block cells with 50% human serum. Neutralizes the human interferon α receptor (IFN-αR) by binding to human interferon α receptor with high affinity and blocking the biological actions of type I interferons. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service