Skip to Content
Merck
All Photos(1)

Key Documents

407294

Sigma-Aldrich

αA-Interferon, Human, Recombinant, E. coli

Synonym(s):

IFNA2A, IFNA2B, IFNA2C, rhIFN-αA

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$641.00

$641.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 EA
$641.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$641.00


Estimated to ship on21 May 2025


Request a Bulk Order

description

Merck USA index - 14, 4990

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥2.8 x 108 units/mg protein
≥5 x 106 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−70°C

Gene Information

human ... IFNA1(3340)

General description

Research area: Immunoand cks

Interferons are cytokines secreted by mammalian cells. They are broadly divided into type I– interferon alpha (IFN-α) and interferon beta (IFN-β), type II – interferon-gamma (IFN-γ), and type III – interferon lambda (IFN-λ) based on their structure, function, and receptor specificity. Recombinant αA interferon is a hydrophobic protein belonging to the IFN-α family of proteins.[1]

Application

Interferon alpha has been used to evaluate its effect on VSV (vesicular stomatitis virus) infection in pancreatic ductal adenocarcinoma (PDA) cell lines and study the relationship between type I IFNsignaling, resistance to VSV infection, and the JAK/STAT (Janus kinase/signal transducers and activators of transcription) signaling pathways in PDA cell lines.[2] It has also been used to study the inhibitory effect of Morbillivirus V Proteins on Interferon Signaling Pathways.[3]

Biochem/physiol Actions

Recombinant, human αA-interferon expressed in E. coli have antiviral, antiproliferative, antineoplastic, and immunomodulating properties. Recombinant interferon αA (IFN-αA) is used as the first line of treatment for hepatitis C.[1] It also displays a great therapeutic potential with chronic viral hepatitis, polycythemia, leukemia, and various other malignancies. It enhances natural killer cell activity, suppresses bone marrow function, and induces apoptotic cell death in non-melanoma skin cancer.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit of interferon per ml of medium will cause a 50% inhibition of VSV-induced cytopathic effect on bovine MDBK cells. Units are determined with respect to the NIH international reference standard for human αA-IFN. Dilute in aqueous buffer containing at least 0.1% BSA or HSA.

Physical form

In PBS containing 0.1% BSA.

Preparation Note

Dilute only in buffers containing protein such as 0.1% BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Fujishima, H., et al. 1998. Int. J. Cancer 76, 423.
Haria, M., and Benfield, P. 1995. Drugs 50, 873.
Familletti, P.C., et al. 1981. Methods Enzymol. 78, 387.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of Soluble Active Interferon ?A in Escherichia coli Periplasm by Fusion with Thermostable Lichenase Using the Domain Insertion Approach
Tyurin AA, et al.
Biochemistry, 83(3), 259-269 (2018)
Morbillivirus V Proteins Exhibit Multiple Mechanisms to Block Type 1 and Type 2 Interferon Signalling Pathways
Senthil K, et al.
PLoS ONE, 8(2), e57063-e57063 null
Resistance of pancreatic cancer cells to oncolytic vesicular stomatitis virus: role of type I interferon signaling
Moerdyk-Schauwecker M, et al.
Virology, 436(1), 221-234 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service