Skip to Content
Merck
All Photos(1)

Key Documents

407255

Sigma-Aldrich

IGFBP-rp1/IGFBP-7, His-Tag, CF, Mouse, Recombinant

Synonym(s):

IGFBP-7

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$662.00

$662.00


Estimated to ship on12 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$662.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$662.00


Estimated to ship on12 May 2025


Request a Bulk Order

Assay

≥97% (SDS-PAGE)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse insulin-like growth factor binding protein-related protein 1 (IGFBP-rp1) consisting of amino acids 28-281 and expressed in S. frugiperda 21 cells with an N-terminal His•Tag. Mouse IGFBP-rp1 cDNA encodes a 281 amino acid precursor protein which contains a putative 25 aa signal peptide and a 256 aa mature region. IGFBP modifies affinities of the binding proteins to IGF.

Biochem/physiol Actions

Functional ELISA Immobilized rmIGFBP-rp1/IGFBP-7 (500 ng/ml) produces 50% optimal binding to rhCCL21/6Ckine at 4-20 ng/ml.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm sterile-filtered PBS.

Reconstitution

Reconstitute to 100 µg/ml in sterile PBS.

Other Notes

Ahmed, S. et al. 2003. Biochem. Biophys. Res. Commun.310, 612.
Nagakubo, D. et al. 2003. J. Immunol.171, 553.
Wilson, HM. et al. 2002. Cell Growth Differ.13, 205.
Degeorges, A. et al. 2000. J. Histochem. Cytochem.48, 747.
Kato, M.V. 2000. Mol. Med.6, 126.
Hwa, V. et al. 1999. Endocr. Rev.20, 761.
Kim, HS. et al. 1997. Proc. Natl. Acad. Sci. USA94,12981.
Kato, M.V. et al. 1996. Oncogene12, 1361.
Oh, Y. et al. 1996. J. Biol. Chem.271, 30322.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service