Skip to Content
Merck
All Photos(1)

Key Documents

4055-OP

Millipore

Ethylenediaminetetraacetic acid disodium salt solution

OmniPur® Grade

Synonym(s):

Ethylenediaminetetraacetic Acid, Disodium Salt, Dihydrate Solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$94.30
500 ML
$307.00

$94.30


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$94.30
500 ML
$307.00

About This Item

UNSPSC Code:
12352107
NACRES:
NA.28

$94.30


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

liquid

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

color

clear colorless

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

General description

Molecular biology grade.

Specifications:
DNase: None detected
pH of solution at 25°C: 7.9-8.1
Protease: None detected
RNase: None detected
Sterility: To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Dual-Responsive Hydrogels for Mercury Ion Detection and Removal from Wastewater.: This study explores the development of dual-responsive hydrogels incorporating ethylenediaminetetraacetic acid (EDTA) for the efficient detection and removal of mercury ions from wastewater, highlighting their potential in environmental biochemistry applications (Diacon et al., 2024).

  • Photocatalytic Hydrogen Evolution Activity of Nitrogen/Fluorine-Codoped Rutile TiO(2).: This article discusses the use of EDTA in the preparation of photocatalytic materials for hydrogen production, emphasizing its role in enhancing photocatalytic efficiency and its relevance in sustainable energy biochemistry (Miyoshi et al., 2023).

  • High-Efficiency Cleaning Technology and Lifespan Prediction for the Ceramic Membrane Treating Secondary Treated Effluent.: This study investigates the application of EDTA in the cleaning processes of ceramic membranes used in wastewater treatment, demonstrating its effectiveness in extending membrane lifespan and improving treatment efficiency (Lv et al., 2023).

  • Reversible Morphology Locking via Metal Infiltration in a Block Copolymer.: The research utilizes EDTA in the reversible locking of block copolymer morphologies through metal infiltration, providing innovative techniques for materials science and biochemistry applications (Ma et al., 2023).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service