Skip to Content
Merck
All Photos(1)

Key Documents

401485

Sigma-Aldrich

IKK-2 Inhibitor, SC-514

InSolution, ≥95%

Synonym(s):

InSolution IKK-2 Inhibitor, SC-514

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$210.00

$210.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 MG
$210.00

About This Item

Empirical Formula (Hill Notation):
C9H8N2OS2
Molecular Weight:
224.30
UNSPSC Code:
12352200
NACRES:
NA.77

$210.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

clearpale yellow

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable, potent, reversible, ATP-competitive and highly selective inhibitor of IKK-2 (IC50 ~3-12 µM for IKK-2 homodimer, IKK-1/IKK-2 heterodimer, and IKK-2) that also displays anti-inflammatory properties. Specificity has been reported using a panel of 31 additional kinases, including IKK isoforms IKK-1, IKK-i, and TBK-1 (IC50 >200 µM). Shown to specifically block NF-κB-dependent gene expression, but not MAP kinase pathways, in stimulated RASF synovial fibroblast cells. Does not inhibit the phosphorylation and activation of the IKK complex.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IKK-2
Product competes with ATP.
Reversible: yes
Target IC50: ~ 3-12 µM for IKK-2 homodimer, IKK-1/IKK-2 heterodimer, and IKK-2

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 25 mM (1 mg/178 µl) solution of IKK-2 Inhibitor, SC-514 (Cat. No. 401479) in DMSO.

Reconstitution

Following initial use, aliquot and refrigerate (4°C).

Other Notes

Baxter, A., et al. 2004 Bioorg. Med. Chem. Lett.14, 2817.
Kishore, N., et al. 2003. J. Biol. Chem.278, 32861.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service