Skip to Content
Merck
All Photos(1)

Key Documents

391333

Millipore

HEPES sodium salt

ULTROL® Grade, ≥99% dry basis (titration), zwitterionic N-substituted aminosulfonic acid buffer

Synonym(s):

HEPES, Sodium Salt, ULTROL® Grade, N-2-Hydroxyethylpiperazine-Nʹ-2-ethanesulfonic Acid, Na

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$238.00
500 G
$555.00
1 KG
$1,240.00

$238.00


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$238.00
500 G
$555.00
1 KG
$1,240.00

About This Item

Empirical Formula (Hill Notation):
C8H17N2O4S · Na
CAS Number:
Molecular Weight:
260.29
MDL number:
UNSPSC Code:
12161704
NACRES:
NA.25

$238.00


Available to ship on02 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥99% dry basis (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

pKa (20 °C)

7.6

solubility

water: 1.0 M

anion traces

chloride (Cl-): <0.01%
sulfate (SO42-): <0.05%

cation traces

heavy metals: <1 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCN1CCN(CC1)CCO

InChI

1S/C8H18N2O4S.Na/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

RDZTWEVXRGYCFV-UHFFFAOYSA-M

General description

A zwitterionic N-substituted aminosulfonic acid buffer(Absorbance: <0.10 at 260 nm, 1.0 mM, H2O). Active in the pH range of 6.0 to 8.5. Has a pKa of 7.6 at 20°C.
Absorbance (1.0 mM, H2O, 260 nm):<0.10.

Application


  • 3,6-O-[N-(2-Aminoethyl)-acetamide-yl]-chitosan exerts antibacterial activity by a membrane damage mechanism.: This study explores the antibacterial mechanisms of a chitosan derivative, which could be related to the buffering capabilities of HEPES in maintaining the stability and exposure of functional groups necessary for antibacterial activity (Yan et al., 2016).

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service