Skip to Content
Merck
All Photos(1)

Key Documents

390602

Sigma-Aldrich

Hydroxyfasudil

A cell-permeable, hydroxylated metabolite of HA 1077 that displays anti-anginal properties.

Synonym(s):

Hydroxyfasudil, ROCK Inhibitor XIV, 1-(1-Hydroxy-5-isoquinolinesulfonyl)homopiperazine, HCl, HA1100

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$247.00

$247.00


Estimated to ship on29 May 2025



Select a Size

Change View
2 MG
$247.00

About This Item

Empirical Formula (Hill Notation):
C14H17N3O3S · xHCl
CAS Number:
Molecular Weight:
307.37 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$247.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

SMILES string

[S](=O)(=O)(N3CCNCCC3)c1c2c([c]([nH]cc2)=O)ccc1.Cl

InChI

1S/C14H17N3O3S.ClH/c18-14-12-3-1-4-13(11(12)5-7-16-14)21(19,20)17-9-2-6-15-8-10-17;/h1,3-5,7,15H,2,6,8-10H2,(H,16,18);1H

InChI key

XWWFOUVDVJGNNG-UHFFFAOYSA-N

General description

A cell-permeable, hydroxylated metabolite of HA 1077 (Fasudil; Cat. No. 371970) that displays anti-anginal properties. Acts as an ATP-competitive and a reversible inhibitor of Rho-kinase (IC50 = 0.9 and 1.8 µM using a peptide and MLC as substrate, respectively) with ~100-fold greater selectivity over MLCK, MRCKβ, and PKC. Reported to inhibit the Rho kinase-mediated contraction both in vitro and in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Rho-kinase
Product competes with ATP.
Reversible: yes
Target IC50: 0.9 and 1.8 µM against Rho-kinase using a peptide and MLC as substrate, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Physical form

A 10 mM (2 mg/582 µl) solution in H₂O.

Other Notes

Ito, K., et al. 2003. J. Physiol.546, 823.
Takemoto, M., et al. 2002. Circulation106, 57.
Shimokawa, H., et al. 1999. Cardiovasc. Res.43, 1029.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service