Skip to Content
Merck
All Photos(1)

Key Documents

385880

Sigma-Aldrich

Hsp25 Kinase Inhibitor

The Hsp25 Kinase Inhibitor controls the biological activity of Hsp25 KInase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Hsp25 Kinase Inhibitor, KKKALNRQLGVAA, MK2 Inhibitor, MAPKAP Kinase-2 Inhibitor, MK2 Inhibitor, KKKALNRQLGVAA, MAPKAP Kinase-2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$280.00

$280.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 MG
$280.00

About This Item

Empirical Formula (Hill Notation):
C61H113N21O16
Molecular Weight:
1396.68
UNSPSC Code:
12352200
NACRES:
NA.77

$280.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

A 13-residue, cell-permeable peptide that acts as a potent and selective inhibitor of mammalian heat-shock protein (Hsp25) kinase. Inhibits Hsp25 kinase competitively with respect to the substrate peptide (Ki = 8.1 µM) and non-competitive with respect to ATP (Ki = 134 µM). [Hsp 25 kinase is also called mitogen-activated protein kinase-activated protein kinase-2 (MK2).]

Biochem/physiol Actions

Cell permeable: yes
Primary Target
(Hsp25) kinase
Product does not compete with ATP.
Reversible: no
Target Ki: 8.1 µM against (Hsp25) kinase

Warning

Toxicity: Standard Handling (A)

Sequence

H-Lys-Lys-Lys-Ala-Leu-Asn-Arg-Gln-Leu-Gly-Val-Ala-Ala-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following recosntitution, aliquot and freeze (-20°C). Stock solutions are stable for up to months at -20°C.

Other Notes

Katsoulidis, E., et al. 2005. Cancer Res.65, 9029.
Hayess, K., and Benndorf, R. 1997. Biochem. Pharmacol. 53, 1239.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service