Skip to Content
Merck
All Photos(1)

Key Documents

382190

Sigma-Aldrich

Histone Lysine Methyltransferase Inhibitor

The Histone Lysine Methyltransferase Inhibitor, also referenced under CAS 935693-62-2, controls the biological activity of Histone Lysine Methyltransferase. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Histone Lysine Methyltransferase Inhibitor, 2-(Hexahydro-4-methyl-1H-1,4-diazepin-1-yl)-6,7-dimethoxy-N-(1-(phenylmethyl)-4-piperidinyl)-4-quinazolinamine, BIX-01294, HMTase Inhibitor, EHMT2/G9a Inhibitor I, EHMT2/G9a Inhibitor I, HMTase Inhibitor, 2-(Hexahydro-4-methyl-1H-1,4-diazepin-1-yl)-6,7-dimethoxy-N-(1-(phenylmethyl)-4-piperidinyl)-4-quinazolinamine, BIX-01294

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38N6O2
CAS Number:
Molecular Weight:
490.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Product 382190 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N2(CCC(CC2)Nc3nc(nc5c3cc(c(c5)OC)OC)N4CCN(CCC4)C)Cc1ccccc1

InChI

1S/C28H38N6O2/c1-32-12-7-13-34(17-16-32)28-30-24-19-26(36-3)25(35-2)18-23(24)27(31-28)29-22-10-14-33(15-11-22)20-21-8-5-4-6-9-21/h4-6,8-9,18-19,22H,7,10-17,20H2,1-3H3,(H,29,30,31)

InChI key

OSXFATOLZGZLSK-UHFFFAOYSA-N

General description

A cell-permeable diazepinyl-quinazolinamine, non-SAM (S-adenosylmethionine) analog-based HMTase (histone methyltransferase) inhibitor that selectively interferes with the G9a-catalyzed H3K9me2 (histone H3 Lys9 dimethylation) modification (IC50 = 1.7 µM) in a reversible manner. It inhibits the GLP-catalyzed H3K9me3 only at much higher concentrartions (IC50 = 38 µM) and exhibits little activity against H3 methylations catalyzed by other HMTases (PRMT1, SET7/9, ESET, SUV39H1). Shown to effectively synergize with Oct3/4 and Klf4 in inducing reprogramming of primary murine fetal NPCs (Neural Progenitor Cells) into iPS (induced Pluripotent Stem) cells without additional viral transduction of Sox2 and c-Myc.
A cell-permeable diazepinyl-quinazolinamine, non-SAM (S-adenosylmethionine) analog-based HMTase (histone methyltransferase) inhibitor that selectively interferes with the G9a-catalyzed H3K9me2 (histone H3 Lys9 dimethylation) modification (IC50 = 1.7 µM) in a reversible manner. It inhibits the GLP-catalyzed H3K9me3 only at much higher concentrations (IC50 = 38 µM) and exhibits little activity against H3 methylations catalyzed by other HMTases (PRMT1, SET7/9, ESET, SUV39H1). Shown to effectively synergize with Oct3/4 and Klf4 in inducing reprogramming of primary murine fetal NPCs (Neural Progenitor Cells) into iPS (induced Pluripotent Stem) cells without additional viral transduction of Sox2 and c-Myc.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Shi, Y., et al. 2008. Cell Stem Cell2, 525.
Kubicek, S., et al. 2007. Mol. Cell25, 473.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service