Skip to Content
Merck
All Photos(1)

Documents

376816

Sigma-Aldrich

Hexokinase II VDAC Binding Domain Peptide, Cell-Permeable

A cell-permeable peptide analog of Hexokinase II VDAC binding domain peptide.

Synonym(s):

Hexokinase II VDAC Binding Domain Peptide, Cell-Permeable, HXK2VBD-cpm, H- RQIKIWFQNRRMKWKK-MIASHLLAYFFTELN-NH₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C188H291N53O40S2
Molecular Weight:
3997.78
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable peptide analog of Hexokinase II VDAC binding domain (HXK2VBD) peptide (Cat. No. 376815). The internalization sequence of the Antennapedia homeoprotein is fused to the N-terminal methionine of the HXK2VBD peptide. Has been shown to cause the complete release of hexokinase II (HXK2) from the mitochondria and subsequent translocation to the cytosol in HeLa cells (100% release at 100 µM). Does not induce Bax translocation or cytochrome c release. Also shown to markedly sensitize cells to Bax-induced cytochrome c release. Also increases the sensitivity of cells to apoptosis in combination with the Bax-dependent apoptosis inducer, Indomethicin (Cat. No. 405268).
A cell-permeable peptide analog of Hexokinase II VDAC binding domain peptide (Cat. No. 376815). The internalization domain of the Antennapedia homeoprotein is fused to the methionine amino terminal. Shown to completely detach and translocate HXK2 from mitochondria to the cytosol in HeLa cells at 100 µM. Does not induce Bax translocation or cytochrome c release when used alone. However, it markedly sensitizes cells to cytochrome c release and to the induction of apoptosis when used in combination with a Bax-dependent apoptosis inducer, Indomethacin (Cat. No. 405268).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Detach and translocate HXK2 from mitochondria to the cytosol
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Met-Ile-Ala-Ser-His-Leu-Leu-Ala-Tyr-Phe-Phe-Thr-Glu-Leu-Asn-NH₂

Other Notes

Pastorino, J.G., et al. 2002. J. Biol. Chem.277, 7610.
Holinger, E.P., et al. 1999. J. Biol. Chem.274, 13298.
Sui, D., and Wilson, J.E. 1997. Arch. Biochem. Biophys.345, 111.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Sun et al.
Redox biology, 39, 101811-101811 (2020-12-29)
Vascular complications of diabetes are a serious challenge in clinical practice, and effective treatments are an unmet clinical need. Acidic fibroblast growth factor (aFGF) has potent anti-oxidative properties and therefore has become a research focus for the treatment of diabetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service