Skip to Content
Merck
All Photos(1)

Key Documents

375115

Sigma-Aldrich

Heparin Cofactor II, Human Plasma

Heparin Cofactor II, Human Plasma, is a native human plasma protein that ihibits thrombin by forming a bimolecular complex in the presence of heparin. It shares homology with antithrombin III

Synonym(s):

Antithrombin BM

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$728.00

$728.00


Estimated to ship on12 May 2025



Select a Size

Change View
100 μG
$728.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$728.00


Estimated to ship on12 May 2025


Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥700 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<2% antigenic ATIII

shipped in

ambient

storage temp.

−20°C

General description

Native heparin cofactor II from human plasma. Inhibits thrombin by forming a bimolecular complex in the presence of heparin. Serine proteinase plasma glycoprotein.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of heparin cofactor II that will inhibit 1 NIH unit of thrombin.

Physical form

In 50% aqueous glycerol.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service