373270
Hec1/Nek2 Mitotic Pathway Inhibitor I, INH1
The Hec1/Nek2 Mitotic Pathway Inhibitor I, INH1, also referenced under CAS 313553-47-8, controls the biological activity of Hec1/Nek2 Mitotic Pathway.
Synonym(s):
Hec1/Nek2 Mitotic Pathway Inhibitor I, INH1, Inhibitor of Nek2/Hec1-1, N-(4-(2,4-Dimethylphenyl)-thiazol-2-yl)-benzamide, INH1, N-(4-(2,4-Dimethylphenyl)-thiazol-2-yl)-benzamide, INH1, Inhibitor of Nek2/Hec1-1
Select a Size
$773.00
Estimated to ship on29 May 2025
Select a Size
About This Item
$773.00
Estimated to ship on29 May 2025
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 50 mg/mL
ethanol: 50 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
[s]1c(nc(c1)c3c(cc(cc3)C)C)NC(=O)c2ccccc2
InChI
1S/C18H16N2OS/c1-12-8-9-15(13(2)10-12)16-11-22-18(19-16)20-17(21)14-6-4-3-5-7-14/h3-11H,1-2H3,(H,19,20,21)
InChI key
JPMOKRWIYQGMJL-UHFFFAOYSA-N
General description
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service