Skip to Content
Merck
All Photos(1)

Documents

371723

Sigma-Aldrich

Anti-Giα-1 and Giα-2-Subunits, C-Terminal (345-354 and 346-355) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... GNAI1(2770)

General description

Anti-Giα-1 and Giα-2-Subunits, C-Terminal (345-354 and 346-355), rabbit polyclonal, recognizes Giα-1 and Giα-2 subunits. It is validated for use in Western blotting.
Protein A purified rabbit polyclonal antibody. Recognizes Giα-1 and Giα-2 subunit proteins.
Recognizes Giα-1 and Giα-2 subunits. Does not cross-react with G3α-3 or Goα subunits.

Immunogen

a synthetic peptide [(C)KNNLKDCGLF] corresponding to amino acids at the C-terminus of mammalian Giα-1 and Giα-2, conjugated to KLH

Application

Immunoblotting (1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

In 140 mM NaCl, 100 mM potassium phosphate, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Does not cross-react with Giα-3 and Goα. The specificity and cross-reactivity were confirmed with lysates from separate cultures of bacteria transfected with the genes for Giα-1, Giα-2, Giα-3, and Goα. Variables associated with assay conditions will dictate the proper working dilution.
Mumby, S.M., and Gilman, A.G. 1991. Methods Enzymol.195, 215.
Goldsmith, P.P., et al. 1987. J. Biol. Chem.262, 14683.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Xu et al.
Infection and immunity, 64(2), 593-599 (1996-02-01)
In previous studies, an in vitro ADP-ribosylation assay was developed to quantitatively evaluate the in vivo ADP-ribosylation of eukaryotic target proteins in intact Chinese hamster ovary (CHO) cells by pertussis toxin (PT). Immunoblot analysis identified the two PT-sensitive target proteins
C Puma et al.
Journal of neurochemistry, 78(5), 960-971 (2001-09-13)
The chemokine IL-8 is known to be synthesized by glial cells in the brain. It has traditionally been shown to have an important role in neuroinflammation but recent evidence indicates that it may also be involved in rapid signaling in
K Takano et al.
Endocrinology, 138(6), 2405-2409 (1997-06-01)
SRIF activates an inwardly rectifying K+ current in human GH-secreting adenoma cells. Activation of this K+ current induces hyperpolarization of the membrane and abolishment of action potential firing. This mechanism is an essential mechanism for SRIF-induced decrease in intracellular Ca2+
Suzy D Carvalho-Bianco et al.
Molecular endocrinology (Baltimore, Md.), 18(7), 1840-1849 (2004-05-08)
Whereas many cardiac symptoms of thyrotoxicosis resemble those of the hyperadrenergic state, circulating catecholamines are reduced or normal in this condition. To test the hypothesis that the thyrotoxic heart is hypersensitive to catechol-amines, we studied beta-adrenergic signaling in a transgenic
Jennifer S Ignatious Raja et al.
The European journal of neuroscience, 39(8), 1245-1255 (2014-01-22)
Intracellular signaling in insect olfactory receptor neurons remains unclear, with both metabotropic and ionotropic components being discussed. Here, we investigated the role of heterotrimeric Go and Gi proteins using a combined behavioral, in vivo and in vitro approach. Specifically, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service