Skip to Content
Merck
All Photos(1)

Key Documents

371706

Sigma-Aldrich

GPR40 Agonist

The GPR40 Agonist, also referenced under CAS 885101-89-3, controls the biological activity of GPR40. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

GPR40 Agonist, 3-(4-(((3-(Phenoxy)phenyl)methyl)amino)phenyl)propanoic acid, FFA1 Agonist I, Free Fatty Acid Receptor 1 Agonist I

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$248.00

$248.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$248.00

About This Item

Empirical Formula (Hill Notation):
C22H21NO3
CAS Number:
Molecular Weight:
347.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$248.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 40 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(Cc2cc(ccc2)Oc3ccccc3)c1ccc(cc1)CCC(=O)O

InChI

1S/C22H21NO3/c24-22(25)14-11-17-9-12-19(13-10-17)23-16-18-5-4-8-21(15-18)26-20-6-2-1-3-7-20/h1-10,12-13,15,23H,11,14,16H2,(H,24,25)

InChI key

DGENZVKCTGIDRZ-UHFFFAOYSA-N

General description

A cell-permeable aminophenylpropanoate that acts as a potent and selective agonist for the G-protein coupled receptor (GPCR) GPR40 (EC50 ~50 nM) with much reduced activity towards family members GPR120 (EC50 ~3.5 µM), GPR41/GPR43 (EC50 >50 µM), as well as a panel of eight other fatty acid and prostaglandin receptors. Shown to induces GPR40-dependent Ca2+ mobilization in HEK-293 cells and potentiate glucose-stimulated insulin secretion in MIN6 cells. Reported to exhibit an in vivo half-life of ≥5.3 hr. in rat (p.o. or i.v.) with a bioavailability of 65% and a clearance of 24.9 ml/min/kg.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

McKeown, S C., et al. 2007. Bioorg. Med. Chem. Lett.17, 1584.
Briscoe, C. P., et al. 2006. Br. J. Pharmacol.148, 619.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service