Skip to Content
Merck
All Photos(2)

Key Documents

370625

Sigma-Aldrich

Guanabenz Acetate

A cell-permeable α2-adrenoceptor agonist that lowers protein misfolding in stressed cells and acts as a proteostatis regulator.

Synonym(s):

Guanabenz Acetate, 1-(2,6-Dichlorobenzylideneamino)guanidine, Acetate, WY-8678

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$276.00

$276.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$276.00

About This Item

Empirical Formula (Hill Notation):
C8H8Cl2N4 · xC2H4O2
CAS Number:
Molecular Weight:
231.08 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$276.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1c(c(ccc1)Cl)\C=N\N=C(\[N+H3])/N.[O-]C(=O)C

InChI

1S/C8H8Cl2N4.C2H4O2/c9-6-2-1-3-7(10)5(6)4-13-14-8(11)12;1-2(3)4/h1-4H,(H4,11,12,14);1H3,(H,3,4)/b13-4+;

InChI key

MCSPBPXATWBACD-GAYQJXMFSA-N

General description

A cell-permeable α2-adrenoceptor agonist that lowers protein misfolding in stressed cells (EC50 ~ 400 nM in tunicamycin treated HeLa cells) and acts as a proteostatis regulator. Shown to selectively bind PPP1R15A/GADD34 and disrupt the PPP1R15A-PP1c complex; sustain eIF2α phosphorylation and increase chaperone availability by attenuating translation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Tsaytler, P., et al. 2011. Science332, 91.
Erb, S., et al. 2000. Neuropsychopharmacology23, 138,
Handley, S.L. and Mithani, S., 1984. Naunyn Schmiedebergs Arch. Pharmacol.327, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service