Skip to Content
Merck
All Photos(1)

Key Documents

369080

Millipore

Guanidine Hydrochloride, Molecular Biology Grade

Powerful protein denaturant.

Synonym(s):

Guanidinium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$222.00
1 KG
$814.00

$222.00


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
100 G
$222.00
1 KG
$814.00

About This Item

Empirical Formula (Hill Notation):
CH5N3 · xHCl
CAS Number:
Molecular Weight:
59.07 (free base basis)
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.25

$222.00


Estimated to ship on01 April 2025


Request a Bulk Order

grade

Molecular Biology

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

ethanol: soluble
water: soluble

absorption

≤0.01 at 280 nm
≤0.05 at 260 nm
≤0.15 at 230 nm

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Cl-].NC(=N)N.[H+]

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Powerful protein denaturant widely used in the purification of proteins and nucleic acids (absorbance: ≤0.05 at 260 nm; ≤0.01 at 280 nm); ≤0.l5 at 230 nm).
Powerful protein denaturant.

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service