Skip to Content
Merck
All Photos(1)

Key Documents

362185

Sigma-Aldrich

N-Glycosidase F, Elizabethkingia meningosepticum

Synonym(s):

N-Glycosidase F, Elizabethkingia meningosepticum, Glycopeptidase F, PNGase F

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Product 362185 is not currently sold in your country. Contact Technical Service

biological source

bacterial (Elizabethkingia meningosepticum)

Quality Level

conjugate

(N-linked)

form

liquid

specific activity

≥20,000 units/mg protein
≥4500 units/mL

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

Proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Native N-Glycosidase F from Elizabethkingia meningosepticum. Catalyzes the hydrolysis of asparagine-linked high mannose, as well as hybrid and complex oligosaccharides from glycoproteins. This enzyme will not cleave oligosaccharides containing α1,3-linked core fucose commonly found on plant glycoproteins. N-Glycosidase F will also fail to cleave if the asparagine to which the oligosaccharide is attached is not peptide-bonded to at least one amino acid residue at both the amino and carboxy termini.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the release of N-linked oligosaccharides from 1.0 nmol denatured ribonuclease B per min at 37°C, pH 7.5. Cleavage is monitored by SDS-PAGE. One unit of N-Glycosidase F is equal to 1 IUB milliunit.

Physical form

In 50 mM NaCl, 20 mM Tris-HCl, 1 mM EDTA, pH 7.5.

Other Notes

Means, R.E. and Desrosiers, R.C. 2000. J. Virol.74, 11181.
Fan, J.Q., and Lee, Y.C. 1997. J. Biol. Chem. 272, 27058.
Tarentino, A.L., and Plummer, T.H. 1994. Methods Enzymol. 230, 44.
Trimble, R.B., and Tarentino, A.L. 1991. J. Biol. Chem. 266, 1646.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service