Skip to Content
Merck
All Photos(2)

Key Documents

361569

Millipore

GSK-3beta Inhibitor XXVI - Calbiochem

Synonym(s):

GSK-3beta Inhibitor XXVI - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18N4O
Molecular Weight:
342.39
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL, light yellow

shipped in

ambient

storage temp.

2-8°C

General description

This product has been discontinued.



A cell-permeable, pyrazolone GSK-3β inhibitor (IC50 = 34 nM, in an enzymatic assay) that is highly selective among a panel of 40 kinases that have been profiled with more than 50 fold selectivity. It exhibits no off-target activity against a panel of 44 human and rodent CNS receptors, channels, and transporters except for some modest activity toward 5HT1a. It demonstrates neuroprotective activity of neurons in an oxidative stress model induced by homocysteic acid (HCA) from 1–20 µM, without observable cytotoxicity, and is superior to SB-216763 Cat. No. 361566) at 5 µM in cell micrographs. In addition, it is shown to attenuate locomotor activity in an chlordiazepoxide/amphetamine-induced hyperactivity mice model in vivo at 150 mg/kg.

A cell-permeable, pyrazolone GSK-3β inhibitor (IC50 = 34 nM, in an enzymatic assay) that is highly selective among a panel of 40 kinases that have been profiled with more than 50 fold selectivity. It exhibits no off-target activity against a panel of 44 human and rodent CNS receptors, channels, and transporters except for some modest activity toward 5HT1a. It demonstrates neuroprotective activity of neurons in an oxidative stress model induced by homocysteic acid (HCA) from 1–20 µM, without observable cytotoxicity, and is superior to SB-216763 Cat. No. 361566) at 5 µM in cell micrographs. In addition, it is shown to attenuate locomotor activity in an chlordiazepoxide/amphetamine-induced hyperactivity mice model in vivo at 150 mg/kg.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen, W., et al. 2011. ChemMedChem6, 1587.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service