Skip to Content
Merck
All Photos(1)

Key Documents

361552

Sigma-Aldrich

GSK-3 Inhibitor IX

InSolution, ≥97%

Synonym(s):

InSolution GSK-3 Inhibitor IX

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$222.00

$222.00


Estimated to ship on23 May 2025



Select a Size

Change View
500 μG
$222.00

About This Item

Empirical Formula (Hill Notation):
C16H10BrN3O2
CAS Number:
Molecular Weight:
356.17
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$222.00


Estimated to ship on23 May 2025


Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Brc1cc2c(cc1)\C(=C3\Nc4c(cccc4)C\3N=O)\C(=O)N2

InChI

1S/C16H10BrN3O2/c17-8-5-6-9-12(7-8)19-16(21)13(9)15-14(20-22)10-3-1-2-4-11(10)18-15/h1-7,14,18H,(H,19,21)/b15-13-

InChI key

WFRDXNRAUPBBTO-SQFISAMPSA-N

General description

A cell-permeable, highly potent, selective, reversible, and ATP-competitive inhibitor of GSK-3α/β (IC50 = 5 nM). Specificity was confirmed using various Cdk′s (IC50 = 83 nM for Cdk5/p25, 300 nM for Cdk2/cyclin A, 320 nM for Cdk1/cyclin B, and 10 µM for Cdk4/cyclin D1), MAP kinases, PKA, PKC isoforms, PKG, CK, and IRTK (IC50 ≥ 10 µM). Inhibition of GSK by BIO has been shown to result in the activation of the Wnt signaling pathway and sustained pluripotency of human and murine embryonic stem cells (ESCs).

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: no
Target IC50: 5 nM against GSK-3α/β

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Physical form

A 10 mM (500 µg/140 µl) solution of GSK-3 Inhibitor IX (Cat. No. 361550) in DMSO.

Other Notes

Polychronopoulos, P., et al. 2004. J. Med. Chem.47, 935.
Sato, N., et al. 2004. Nat. Med.10, 55.
Meijer, L., et al. 2003. Chem. Biol.10, 1255.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service