Skip to Content
Merck
All Photos(1)

Key Documents

344145

Sigma-Aldrich

FPTase, Rat, Recombinant, E. coli

A heterodimeric enzyme that catalyzes the transfer of a 15-carbon isoprenoid group to a variety of cellular proteins including Ras.

Synonym(s):

Farnesyl Proteintransferase, FTase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Product 344145 is not currently sold in your country. Contact Technical Service

recombinant

expressed in E. coli

Quality Level

Assay

≥85% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

M.W. α-subunit: ~48,000; β-subunit: ~46,000
Recombinant, rat FPTase expressed in E. coli. A heterodimeric enzyme that catalyzes the transfer of a 15-carbon isoprenoid group to a C-terminal cysteine in a variety of cellular proteins, including Ras. Recognizes the CaaX motif where the farnesylated cysteine (C) is followed by two aliphatic amino acids (aa) and either leucine or serine (X). Expressed in E. coli as an untagged recombinant protein.

Biochem/physiol Actions

1 pmol of FPTase will transfer 1 pmol of farnesyl to H-Ras in 15 min at 37°C.

Warning

Toxicity: Standard Handling (A)

Physical form

In 25 mM HEPES, pH 7.2, 40 mM NaCl, 5µM ZnCl2, and 1mM TECP

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Hightower, K.E., et al. 2001. Biochem. J.360, 625.
Zimmerman, K.K., et al. 1998. Protein Expr. Purif.14, 395.
Reiss, Y., et al. 1990. Cell62, 81.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seol Hwa Shin et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(23), e2102414-e2102414 (2021-10-20)
Human epidermal growth factor receptor 2 (HER2) is overexpressed in breast and gastric cancers and this causes poor clinical outcomes. Although both T-DM1 and Enhertu are approved as an HER2-targeting antibody-drug conjugate (ADC), the effects of these drugs are still

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service