Skip to Content
Merck
All Photos(1)

Key Documents

344039

Sigma-Aldrich

cFMS Receptor Inhibitor IV

The cFMS Receptor Inhibitor IV, also referenced under CAS 959626-45-0, controls the biological activity of cFMS. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

cFMS Receptor Inhibitor IV, 5-Cyano-N-(2,5-di(piperidin-1-yl)phenyl)furan-2-carboxamide, CSF-1 Receptor Inhibitor IV

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$376.00

$376.00


Estimated to ship on30 May 2025



Select a Size

Change View
5 MG
$376.00

About This Item

Empirical Formula (Hill Notation):
C22H26N4O2
CAS Number:
Molecular Weight:
378.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$376.00


Estimated to ship on30 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N4(CCCCC4)c1c(cc(cc1)N3CCCCC3)NC(=O)c2[o]c(cc2)C#N

InChI

1S/C22H26N4O2/c23-16-18-8-10-21(28-18)22(27)24-19-15-17(25-11-3-1-4-12-25)7-9-20(19)26-13-5-2-6-14-26/h7-10,15H,1-6,11-14H2,(H,24,27)

InChI key

FQGXFVDABCBEFW-UHFFFAOYSA-N

General description

A cell-permeable furan carboxamide compound that acts as a potent, reversible, and ATP-competitive cFMS/CSF-1R inhibitor that blocks cFMS autophosphorylation both in CSF-1/M-CSF-stimulated HEK-293 cells (IC50 = 76 nM) and in cell-free in vitro kinase assays (IC50 = 17 nM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Patch, R.J., et al. 2007. Bioorg. Med. Chem. Lett.17, 6070.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service